Learn More
Selleck Chemical LLC MF-438
SDP
Catalog No. 5000002558
Encompass_Preferred
MF-438 is a potent and orally bioavailable stearoyl-CoA desaturase 1 (SCD1) inhibitor with IC50 of 2 3 nM for rSCD1
Catalog No. 50-000-02558 Supplier Selleck Chemical LLC Supplier No. S68425MG
MF-438 is a potent and orally bioavailable stearoyl-CoA desaturase 1 (SCD1) inhibitor with IC50 of 2 3 nM for rSCD1