Learn More
Medchemexpress LLC MF-766 10mg | 1050656-06-8 | 478.46 | C27H21F3N2O3 | 10 MG
SDP
Catalog No. 5000229617
Encompass_Preferred
MF-766 is a potent, selective, and orally active antagonist of the prostaglandin E2 EP4 receptor, used as a research tool in inflammation, immunology, and oncology studies. It shows sub-nanomolar binding affinity and low-nanomolar functional inhibition in cell-based assays.
  • Potent EP4 antagonist with Ki = 0.23 nM and IC50 ≈ 1.4 nM.
  • Suitable for in vitro and in vivo studies; soluble in DMSO (50 mg/mL) and formulatable in 10% DMSO/90% corn oil.
  • High chemical purity (≈99.8%) for consistent experimental results.
  • Solid, white to off-white powder with recommended powder storage at -20°C for long-term stability.
  • Available in small research sizes and as a 10 mM DMSO solution for convenience.
Catalog No. 50-002-29617 Supplier Medchemexpress LLC Supplier No. HY11548710MG
MF-766 is a potent, selective, and orally active antagonist of the prostaglandin E2 EP4 receptor, used as a research tool in inflammation, immunology, and oncology studies. It shows sub-nanomolar binding affinity and low-nanomolar functional inhibition in cell-based assays.
  • Potent EP4 antagonist with Ki = 0.23 nM and IC50 ≈ 1.4 nM.
  • Suitable for in vitro and in vivo studies; soluble in DMSO (50 mg/mL) and formulatable in 10% DMSO/90% corn oil.
  • High chemical purity (≈99.8%) for consistent experimental results.
  • Solid, white to off-white powder with recommended powder storage at -20°C for long-term stability.
  • Available in small research sizes and as a 10 mM DMSO solution for convenience.