Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MF-766 10mM 1mL | 1050656-06-8 | 478.46 | C27H21F3N2O3 | 1 ML
SDP

Catalog No. 5000228166
Encompass_Preferred

MF-766 is a potent, selective EP4 antagonist provided for preclinical research. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and as solid quantities for assay and in vivo studies.

  • Supplied as 10 mM solution in DMSO, 1 mL.
  • Also available as solids (5 mg-500 mg) for custom preparations.
  • High purity suitable for research (99.83%).
  • Molecular weight 478.46, formula C27H21F3N2O3.
  • Recommended solvent for stocks: DMSO; in vivo vehicle example: 10% DMSO in corn oil.

Catalog No. 50-002-28166 Supplier Medchemexpress LLC Supplier No. HY11548710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MF-766 is a potent, selective EP4 antagonist provided for preclinical research. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and as solid quantities for assay and in vivo studies.

  • Supplied as 10 mM solution in DMSO, 1 mL.
  • Also available as solids (5 mg-500 mg) for custom preparations.
  • High purity suitable for research (99.83%).
  • Molecular weight 478.46, formula C27H21F3N2O3.
  • Recommended solvent for stocks: DMSO; in vivo vehicle example: 10% DMSO in corn oil.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.