Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MF-766 50mg | 1050656-06-8 | 478.46 g/mol | C27H21F3N2O3 | 50 MG
SDP

Catalog No. 5000228678
Encompass_Preferred

MF-766 is a potent, selective, orally active EP4 receptor antagonist provided for research use as a solid compound. It is supplied with supporting analytical data and safety documentation to aid experimental use.

  • High purity: 99.83%.
  • Available in solid pack sizes, including 50 mg.
  • Molecular formula C27H21F3N2O3; molecular weight 478.46 g/mol.
  • CAS number 1050656-06-8 for unambiguous identification.
  • Reported EP4 antagonism (Ki ≈ 0.23 nM) suitable for in vitro and in vivo studies.
  • Comes with SDS and product data for handling and storage guidance.

Catalog No. 50-002-28678 Supplier Medchemexpress LLC Supplier No. HY11548750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MF-766 is a potent, selective, orally active EP4 receptor antagonist provided for research use as a solid compound. It is supplied with supporting analytical data and safety documentation to aid experimental use.

  • High purity: 99.83%.
  • Available in solid pack sizes, including 50 mg.
  • Molecular formula C27H21F3N2O3; molecular weight 478.46 g/mol.
  • CAS number 1050656-06-8 for unambiguous identification.
  • Reported EP4 antagonism (Ki ≈ 0.23 nM) suitable for in vitro and in vivo studies.
  • Comes with SDS and product data for handling and storage guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.