Learn More
Medchemexpress LLC MF-766 5mg | 1050656-06-8 | 5 MG
SDP
Catalog No. 5000229221
Encompass_Preferred
MF-766 is a potent, selective, orally active EP4 receptor antagonist used in preclinical research on cancer and inflammatory pathways. The compound is supplied as a white to off-white solid with documented purity, molecular weight, and solubility data to support experimental use.
  • Potent EP4 antagonist with low nanomolar activity in functional assays.
  • High purity suitable for research-grade applications.
  • White to off-white solid for easy handling and storage.
  • High solubility in DMSO for stock solution preparation.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-29221 Supplier Medchemexpress LLC Supplier No. HY1154875MG
MF-766 is a potent, selective, orally active EP4 receptor antagonist used in preclinical research on cancer and inflammatory pathways. The compound is supplied as a white to off-white solid with documented purity, molecular weight, and solubility data to support experimental use.
  • Potent EP4 antagonist with low nanomolar activity in functional assays.
  • High purity suitable for research-grade applications.
  • White to off-white solid for easy handling and storage.
  • High solubility in DMSO for stock solution preparation.
  • Stable when stored under recommended conditions.