Learn More
Medchemexpress LLC MF-DH-300 | 2673403-86-4 | 99.5% | 330.38 | 100 MG
SDP
Catalog No. 5000364868
Encompass_Preferred
MF-DH-300 is a 15-PGDH inhibitor used in research for muscle disorders like spinal muscular atrophy (SMA). 15-PGDH (15-Hydroxyprostaglandin dehydrogenase) acts as a tumor suppressor in various cancers by metabolizing intracellular PGE2, thus suppressing PGE2 signaling. This product is a solid, white to off-white in color, with a molecular formula of C20H18N4O. It is intended for research use only.
  • Applicable to research of muscle disorders such as spinal muscular atrophy
  • Inhibitor of 15-PGDH
  • Solid with a white to off-white color
  • Molecular formula of C20H18N4O
  • Intended for research use only
Catalog No. 50-003-64868 Supplier Medchemexpress LLC Supplier No. 100MGHY176803
MF-DH-300 is a 15-PGDH inhibitor used in research for muscle disorders like spinal muscular atrophy (SMA). 15-PGDH (15-Hydroxyprostaglandin dehydrogenase) acts as a tumor suppressor in various cancers by metabolizing intracellular PGE2, thus suppressing PGE2 signaling. This product is a solid, white to off-white in color, with a molecular formula of C20H18N4O. It is intended for research use only.
  • Applicable to research of muscle disorders such as spinal muscular atrophy
  • Inhibitor of 15-PGDH
  • Solid with a white to off-white color
  • Molecular formula of C20H18N4O
  • Intended for research use only