Learn More
Medchemexpress LLC MF-DH-300 | 2673403-86-4 | 99.5% | 330.38 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY176803
Encompass_Preferred
MF-DH-300 is an inhibitor of 15-PGDH (15-Hydroxyprostaglandin dehydrogenase), designed for research applications. It is particularly relevant for studies involving muscle disorders, such as spinal muscular atrophy (SMA). This compound is available in solid and solution forms for various experimental needs.
  • Inhibits 15-PGDH activity.
  • Applicable to muscle disorder research.
  • Available in solid or solution forms.
  • High purity.
  • Provides potential for studying spinal muscular atrophy.
Catalog No. 50-003-65873