Learn More
Medchemexpress LLC MF-DH-300 | 2673403-86-4 | 99.5% | 330.38 | 5 MG
SDP
Catalog No. 5000393353
Encompass_Preferred
MF-DH-300 is a 15-PGDH inhibitor applicable for research into muscle disorders, such as spinal muscular atrophy (SMA). 15-PGDH (15-Hydroxyprostaglandin dehydrogenase) acts as a tumor suppressor, and its decreased expression is linked to increased tumorigenesis. This inhibitor metabolizes intracellular PGE2, preventing it from binding to EP receptors and thereby suppressing PGE2 signaling.
  • Can be used for research on muscle disorders
  • Inhibits 15-PGDH enzyme
  • Suppresses PGE2 signaling
  • High purity of 99.53%
  • Solid appearance
Catalog No. 50-003-93353 Supplier Medchemexpress LLC Supplier No. 5MGHY176803
MF-DH-300 is a 15-PGDH inhibitor applicable for research into muscle disorders, such as spinal muscular atrophy (SMA). 15-PGDH (15-Hydroxyprostaglandin dehydrogenase) acts as a tumor suppressor, and its decreased expression is linked to increased tumorigenesis. This inhibitor metabolizes intracellular PGE2, preventing it from binding to EP receptors and thereby suppressing PGE2 signaling.
  • Can be used for research on muscle disorders
  • Inhibits 15-PGDH enzyme
  • Suppresses PGE2 signaling
  • High purity of 99.53%
  • Solid appearance