Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Single-Use Precision Pump Tubing, Gamma-Irradiated C-Flex, L/S 16; 25 ft
Catalog No. NC1496609
Encompass
Masterflex gamma-irradiated pump tubing protects your sensitive processes and products from accidental contamination while saving time! Gamma-irradiated tubing helps you avoid costly delays that can be associated with outsourced gamma irradiation services. Tubing is double bagged prior to undergoing the gamma irradiation sterilization process, and all sealed tubing is gamma irradiated at 32.5 kGy to offer optimal microbial control in a critical fluid pathway. Tubing boxes include gamma radiation-sensitive labels to confirm treatment. Ideal for pharmaceutical and biopharmaceutical processing, sensitive research, single-use systems, and more. Available formulations include: Platinum-Cured Silicone PharMed BPT C-Flex (001)
Catalog No. NC1496609 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9612316
Masterflex gamma-irradiated pump tubing protects your sensitive processes and products from accidental contamination while saving time! Gamma-irradiated tubing helps you avoid costly delays that can be associated with outsourced gamma irradiation services. Tubing is double bagged prior to undergoing the gamma irradiation sterilization process, and all sealed tubing is gamma irradiated at 32.5 kGy to offer optimal microbial control in a critical fluid pathway. Tubing boxes include gamma radiation-sensitive labels to confirm treatment. Ideal for pharmaceutical and biopharmaceutical processing, sensitive research, single-use systems, and more. Available formulations include: Platinum-Cured Silicone PharMed BPT C-Flex (001)