Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P Variable-Speed Air-Powered Drive, 100 to 650 rpm
Catalog No. NC1651571
Encompass
Use this variable-speed air-powered drive when electrical power may be unsafe or impractical. For safe operation, carefully ground to protect from static electricity.
Catalog No. NC1651571 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0758930
Use this variable-speed air-powered drive when electrical power may be unsafe or impractical. For safe operation, carefully ground to protect from static electricity.