Learn More
Sigma Aldrich Fine Chemicals Biosciences Mf MilliporeR Membrane fil
Catalog No. 502904532
Encompass
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made Mf-Millipore membrane filters one of the most widely used membranes in analytical and research applications. Mf-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard Mf-Millipore filters.features Benefits - Versatile filter for biological and environmental monitoring applications - Available in a range of pore sizes colored black or white with or without a gridded surface - Compatible with ethylene oxide gamma irradiation and autoclave sterilization methods
Catalog No. 50-290-4532 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HAWG047500
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made Mf-Millipore membrane filters one of the most widely used membranes in analytical and research applications. Mf-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard Mf-Millipore filters.features Benefits - Versatile filter for biological and environmental monitoring applications - Available in a range of pore sizes colored black or white with or without a gridded surface - Compatible with ethylene oxide gamma irradiation and autoclave sterilization methods