Learn More
Azzota Corp Microporous Filter, NYLON, Diameter: 47mm, Pore Size: 0.45um, 100/pk
Catalog No. NC2328569
Encompass_Preferred
Microporous Filter, NYLON, Diameter: 47mm, Pore Size: 0.45um, 100/pk.
Catalog No. NC2328569 Supplier Azzota Corp Supplier No. BF470145
Microporous Filter, NYLON, Diameter: 47mm, Pore Size: 0.45um, 100/pk.