Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex B/T PerfectPosition™ Pump Tubing, Platinum-Cured Silicone, B/T 87; 10 ft
Catalog No. NC0701689
Encompass
Masterflex B/T pump tubing is manufactured to extremely close tolerances that match our B/T pump heads, ensuring accurate, repeatable flow and long tubing life. Our pump tubing is factory-tested and optically inspected to provide the best performance from your peristaltic pump.
Catalog No. NC0701689 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9651087/DEL
Masterflex B/T pump tubing is manufactured to extremely close tolerances that match our B/T pump heads, ensuring accurate, repeatable flow and long tubing life. Our pump tubing is factory-tested and optically inspected to provide the best performance from your peristaltic pump.