Learn More
Aviva Systems Biology Corp MFAP2 Antibody - N-terminal region (ARP56961_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56961_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MFAP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MFAP2. Alternate names: MAGP, MAGP-1, MAGP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-62044