Learn More
AdipoGen MFAP4 human rec
SDP
Catalog No. 502563017
Encompass_Preferred
Protein. Human MFAP4 aa 1-255 is fused at the C-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. Microfibril-associated protein 4 MFAP4 was initially identified as a gene commonly deleted in the contiguous Smith-Magenis syndrome SMS gene. MFAP4 was identified as growth factor to support expansion of hematopoietic stem cells in culture. It is involved in calcium-dependent cell adhesion and intercellular interactions.
Catalog No. 50-256-3017 Supplier AdipoGen Supplier No. AG40A0072C050
Protein. Human MFAP4 aa 1-255 is fused at the C-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. Microfibril-associated protein 4 MFAP4 was initially identified as a gene commonly deleted in the contiguous Smith-Magenis syndrome SMS gene. MFAP4 was identified as growth factor to support expansion of hematopoietic stem cells in culture. It is involved in calcium-dependent cell adhesion and intercellular interactions.