Learn More
MOLTOX Molecular Toxicology Inc mFC Agar, 50 mm Plate, 10 - 12 mL/plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948659
Encompass_Preferred
mFC Agar is used with Rosolic Acid to cultivate and enumerate fecal coliforms by membrane filtration at elevated temperatures. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-659 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S206.140
mFC Agar is used with Rosolic Acid to cultivate and enumerate fecal coliforms by membrane filtration at elevated temperatures. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.