Learn More
MOLTOX Molecular Toxicology Inc mFC Broth, 10.5 mL/vial, 10 vials/box, Store in Refrigerator
SDP
Catalog No. 50948788
Encompass_Preferred
mFC Broth is used with Rosolic Acid to cultivate and enumerate fecal coliforms via membrane filtration technique at elevated temperatures. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-788 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26635.057
mFC Broth is used with Rosolic Acid to cultivate and enumerate fecal coliforms via membrane filtration technique at elevated temperatures. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.