Learn More
MFG Tray Chemical-Resistant Multipurpose Trays and Pans
Shop All Molded Fiber Glass Tray Company ProductsDescription
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
- Reinforced white thermoset polyester composite construction Resistant to most detergents, mild acid and alkaline solutions, corrosive salts and many reagents
- Withstand temperatures from -51° to +121°C (-60° to +250°F) continuous exposure, or 149°C (300°F) intermittent
- High load-bearing capacity
- Excellent dimensional stability Easily steam cleaned
Order Info
Available in 1 in. deep (2.5cm) trays, shallow pans from 1 1/4 to 2 3/4 in. deep (3 to 7cm), and ventilated stacking pans from 2 3/4 to 4 1/8 in. deep (7 to 10.5cm).
Specifications
Specifications
| Dimensions (L x W x H) | 25.25 x 19.25 x 1.125 in. |
| Color | White |
| Height (Metric) | 2.75 cm |
| Length (Metric) | 64 cm |
| Material | Fiberglass |
| Type | Multi-purpose Tray and Shallow Pan |
| Width (Metric) | 49 cm |
| For Use With (Application) | Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills |
| Length (English) | 25.25 in. |
| Height (English) | 1.125 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.