Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFG Tray Safety Tray for Reagents

Catalog No. 152392 Shop All Molded Fiber Glass Tray Company Products
Change view
Click to view available options
Dimensions (L x W x H):
30 x 20 x 2 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
15-239-2 30 x 20 x 2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-239-2 Supplier MFG Tray Supplier No. 6081031537L15

missing translation for 'validMainframeMsg'

Lightweight tray resists acids, mild alkalis, and most solvents

Lightweight tray resists acids, mild alkalis, and most solvents

  • Rounded edge contains spills
  • Won't corrode or warp
  • Strong enough to carry several bottles
  • Excellent dimensional stability
  • Clean with steam or heat sterilization

Order Info

Outside dimensions: 30L x 20W x 2 in. H (76 x 51 x 5.1cm)

Compliance

Meets OSHA requirements.

Specifications

Dimensions (L x W x H) 30 x 20 x 2 in.
Height (Metric) 5.08 cm
Length (Metric) 76.2 cm
Material Fiberglass
Type Reagent Tray
Width (Metric) 51 cm
For Use With (Application) Lightweight tray resists Acids, mild alkalis and most solvents
Length (English) 30 in.
Height (English) 2 in.
Shape Rectangular
Width (English) 20 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.