Learn More
MSC MFG Tray Hopper Front Bin, Fiberglass-Reinforced Composite, 12 x 24 x 8 in, Stack-N-Nest
Catalog No. 5000514021
Encompass
This bin is designed with a hopper front style and a rectangular shape, crafted from durable fiberglass-reinforced composite material. It offers practical storage solutions for various industrial and laboratory applications, featuring stackable and nestable capabilities for efficient space utilization.
  • Hopper front style for easy access to contents.
  • Rectangular shape for versatile placement.
  • Constructed from fiberglass-reinforced composite for durability.
  • Stackable design allows for vertical storage to save space.
  • Nestable when empty for compact storage.
  • Overall dimensions are 12 in wide, 24 in long, and 8 in high.
  • Inside dimensions are 7-5/8 in high, 7-5/8 in wide, and 18-7/8 in long.
  • Load capacity of 100 lb.
Catalog No. 50-005-14021 Supplier MSC Supplier No. 48757272
This bin is designed with a hopper front style and a rectangular shape, crafted from durable fiberglass-reinforced composite material. It offers practical storage solutions for various industrial and laboratory applications, featuring stackable and nestable capabilities for efficient space utilization.
  • Hopper front style for easy access to contents.
  • Rectangular shape for versatile placement.
  • Constructed from fiberglass-reinforced composite for durability.
  • Stackable design allows for vertical storage to save space.
  • Nestable when empty for compact storage.
  • Overall dimensions are 12 in wide, 24 in long, and 8 in high.
  • Inside dimensions are 7-5/8 in high, 7-5/8 in wide, and 18-7/8 in long.
  • Load capacity of 100 lb.