Learn More
MSC MFG Tray Stacking Tote, ESD, Molded Fiberglass, 23-3/8 x 12 x 4-3/8 in, Black
Catalog No. 5000533065
Encompass
These ESD-safe stacking containers offer optimal product protection with exceptional weight-carrying capability and superior physical properties. They resist bending or sagging under heavy loads and integrate into automated systems. They operate from -60° to 250°F (-51° to 121°C) and resist cutting oils, greases, solvents, mild acids, and alkaline solutions (pH 3.0-10.0). Cleaning is easy with hot water or steam.
  • Optimal product protection
  • High weight-carrying capability
  • Superior to thermoplastic and corrugated products
  • Resists bending and sagging
  • Integrates into automated applications
  • Complete electrostatic discharge protection
  • Functions from -60° to 250°F (-51° to 121°C)
  • Resistant to oils, greases, solvents, mild acids, and alkaline solutions (pH 3.0-10.0)
  • Easy to clean with hot water or steam
Catalog No. 50-005-33065 Supplier MSC Supplier No. 06900765
These ESD-safe stacking containers offer optimal product protection with exceptional weight-carrying capability and superior physical properties. They resist bending or sagging under heavy loads and integrate into automated systems. They operate from -60° to 250°F (-51° to 121°C) and resist cutting oils, greases, solvents, mild acids, and alkaline solutions (pH 3.0-10.0). Cleaning is easy with hot water or steam.
  • Optimal product protection
  • High weight-carrying capability
  • Superior to thermoplastic and corrugated products
  • Resists bending and sagging
  • Integrates into automated applications
  • Complete electrostatic discharge protection
  • Functions from -60° to 250°F (-51° to 121°C)
  • Resistant to oils, greases, solvents, mild acids, and alkaline solutions (pH 3.0-10.0)
  • Easy to clean with hot water or steam