Learn More
MSC MFG Tray Storage Tote, Fiberglass, 300 lb Capacity, Stacking, Nesting
Catalog No. 5000533511
Encompass
This fiberglass storage tote is designed for industrial use, offering a 300 lb capacity. It features both stacking and nesting capabilities for efficient storage and organization.
  • Offers a 300 lb load capacity
  • Constructed from durable fiberglass material
  • Features stacking capability for efficient vertical storage
  • Allows nesting when empty to save space
  • Withstands temperatures from -60 to 250 degrees F
Catalog No. 50-005-33511 Supplier MSC Supplier No. 00343624
This fiberglass storage tote is designed for industrial use, offering a 300 lb capacity. It features both stacking and nesting capabilities for efficient storage and organization.
  • Offers a 300 lb load capacity
  • Constructed from durable fiberglass material
  • Features stacking capability for efficient vertical storage
  • Allows nesting when empty to save space
  • Withstands temperatures from -60 to 250 degrees F