Learn More
MSC MFG Tray Tote Container, 200 Lb Capacity, Fiberglass, Stacking, Green
Catalog No. 5000533547
Encompass
The MFG Tray fiberglass tote container offers a robust solution for industrial storage and organization. Designed for durability, this stacking tote features a a 200 lb load capacity and is made from strong fiberglass material, suitable for a wide range of applications.
  • Provides 200 lb load capacity for heavy-duty storage.
  • Stacking design allows for efficient use of space.
  • Constructed from durable fiberglass material.
  • Withstands temperatures from -60 to 250 degrees Fahrenheit.
  • Features a vibrant green color for easy identification.
  • Measures 23.4 inches long by 12 inches wide by 3.1 inches high.
Catalog No. 50-005-33547 Supplier MSC Supplier No. 00343814
The MFG Tray fiberglass tote container offers a robust solution for industrial storage and organization. Designed for durability, this stacking tote features a a 200 lb load capacity and is made from strong fiberglass material, suitable for a wide range of applications.
  • Provides 200 lb load capacity for heavy-duty storage.
  • Stacking design allows for efficient use of space.
  • Constructed from durable fiberglass material.
  • Withstands temperatures from -60 to 250 degrees Fahrenheit.
  • Features a vibrant green color for easy identification.
  • Measures 23.4 inches long by 12 inches wide by 3.1 inches high.