Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4240MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: EDM45. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Normal breast or breast carcinoma. Cellular Staining: Cell Surface & Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Delipidated human milk fat globule membrane preparation. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 45kDa.Chromosome: 15q25. Gene Symbol: MFGE8. Gene ID: 4240. Ensemble: ENSG00000140545. Uniprot: NP_001297250.
Catalog No. 50-188-4931