Learn More
Aviva Systems Biology Corp MELTF Antibody - C-terminal region (ARP60335_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60335_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MFI2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human MFI2. Alternate names: CD228, FLJ38863, MAP97, MGC4856, MTF1ApplicationIF, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-010