Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Easy-Load Head for High-Performance Precision Tubing, PARA Housing, SS Rotor
Catalog No. NC1715060
Encompass
This Easy-Load pump head accepts high-performance precision tubing¿ ensures optimal performance from your Masterflex pump. Select a pump head for one of the following precision tubing sizes: L/S 15 or L/S 24.
Catalog No. NC1715060 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0751412
This Easy-Load pump head accepts high-performance precision tubing¿ ensures optimal performance from your Masterflex pump. Select a pump head for one of the following precision tubing sizes: L/S 15 or L/S 24.