Learn More
Accuflow Systems MFLEX SILICONE TUB. #13 25FT
Catalog No. 501108828
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex platinum-cured silicone tubing, L/S 13, 25 ft. Ensure optimal performance from your Masterflex pump. Slightly greater clarity than peroxide-cured tubing; smooth surfaces; lower protein bind
Catalog No. 50-110-8828 Supplier Accuflow Systems Supplier No. 9641013
Manufactured by Cole Parmer and sold by Accuflow. Masterflex platinum-cured silicone tubing, L/S 13, 25 ft. Ensure optimal performance from your Masterflex pump. Slightly greater clarity than peroxide-cured tubing; smooth surfaces; lower protein bind