Learn More
Accuflow Systems MFLEX SILICONE TUB. #15 25FT
Catalog No. 501109134
Encompass_Preferred
Manufactured by Cole Parmer & sold by Accuflow. Masterflex platinum-cured silicone tubing, L/S 15, 25 ft. Slightly greater clarity than peroxide-cured tubing Smooth surfaces; lower protein binding levels Fewer potential leachables Ideal fo
Catalog No. 50-110-9134 Supplier Accuflow Systems Supplier No. 9641015
Manufactured by Cole Parmer & sold by Accuflow. Masterflex platinum-cured silicone tubing, L/S 15, 25 ft. Slightly greater clarity than peroxide-cured tubing Smooth surfaces; lower protein binding levels Fewer potential leachables Ideal fo