Learn More
Accuflow Systems MFLEX SILICONE TUB. #25 25FT
Catalog No. 501109139
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex platinum-cured silicone tubing, L/S 25, 25 ft. Ensure optimal performance from your Masterflex pump Slightly greater clarity than peroxide-cured tubing Smooth surfaces; lower pr
This item is not returnable. View return policy
Catalog No. 50-110-9139 Supplier Accuflow Systems Supplier No. 9641025
Manufactured by Cole Parmer and sold by Accuflow. Masterflex platinum-cured silicone tubing, L/S 25, 25 ft. Ensure optimal performance from your Masterflex pump Slightly greater clarity than peroxide-cured tubing Smooth surfaces; lower pr