Learn More
Accuflow Systems MFLEX SILICONE TUB. #36 25FT
Catalog No. 501109141
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex platinum-cured silicone tubing, L/S 36, 25 ft. Ensure optimal performance from your Masterflex pump Slightly greater clarity than peroxide-cured tubing Smooth surfaces; lower pr
Catalog No. 50-110-9141 Supplier Accuflow Systems Supplier No. 9641036
Manufactured by Cole Parmer and sold by Accuflow. Masterflex platinum-cured silicone tubing, L/S 36, 25 ft. Ensure optimal performance from your Masterflex pump Slightly greater clarity than peroxide-cured tubing Smooth surfaces; lower pr