Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Precision Pump Tubing, Peroxide-Cured Silicone, L/S 13; 25 ft
Catalog No. NC9602878
Encompass
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent
Catalog No. NC9602878 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9640013
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent