Learn More
Accuflow Systems MFLEX TUBING CHEM BIO #26 50FT
Catalog No. 501109183
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex I/P, Chem-Durance Bio tubing, #26, 50' Applications: Laboratory, pharmaceutical processing, and biotech applications for pump lines. Characteristics: Excellent chemical resistance. Excellent li
Catalog No. 50-110-9183 Supplier Accuflow Systems Supplier No. 0644226
Manufactured by Cole Parmer and sold by Accuflow. Masterflex I/P, Chem-Durance Bio tubing, #26, 50' Applications: Laboratory, pharmaceutical processing, and biotech applications for pump lines. Characteristics: Excellent chemical resistance. Excellent li