Learn More
Abcam mFluor™ Red 700 Conjugation Kit, 100UG
Catalog No. 502700610
Encompass_Preferred
mFluor™ Red 700 conjugation / labeling in < 4 hrs with 30 secs hands-on time using mFluor™ Red 700 Conjugation Kit ab312807. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-0610 Supplier Abcam Supplier No. AB312807100UG
mFluor™ Red 700 conjugation / labeling in < 4 hrs with 30 secs hands-on time using mFluor™ Red 700 Conjugation Kit ab312807. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement