Learn More
Abcam mFluor™ Red 780 Conjugation Kit, 100UG
Catalog No. 502700609
Encompass_Preferred
mFluor™ Red 780 conjugation / labeling in < 4 hrs with 30 secs hands-on time using mFluor™ Red 780 Conjugation Kit ab312808. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-0609 Supplier Abcam Supplier No. AB312808100UG
mFluor™ Red 780 conjugation / labeling in < 4 hrs with 30 secs hands-on time using mFluor™ Red 780 Conjugation Kit ab312808. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement