Learn More
Abcam mFluor™ Violet 450 Conjugation Kit, 100UG
Catalog No. NC2589123
Encompass_Preferred
mFluor™ Violet 450 conjugation / labeling in < 4 hrs with 30 secs hands-on time using mFluor™ Violet 450 Conjugation Kit ab312804. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2589123 Supplier Abcam Supplier No. AB312804100UG
mFluor™ Violet 450 conjugation / labeling in < 4 hrs with 30 secs hands-on time using mFluor™ Violet 450 Conjugation Kit ab312804. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement