Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P Precision Pump Tubing, Gore™ STA-PURE™ PCS-Series, I/P 82; 24""/Pk
Catalog No. NC1460528
Encompass
Featuring a long pump life and chemically compatible to a wide range of chemicals, genuine Masterflex tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment. Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility. Made by a global market leader in range of formulations for most extensive chemical compatibility.
Catalog No. NC1460528 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9624182
Featuring a long pump life and chemically compatible to a wide range of chemicals, genuine Masterflex tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment. Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility. Made by a global market leader in range of formulations for most extensive chemical compatibility.