Learn More
Aviva Systems Biology Corp MFN1 Antibody - middle region (ARP57702_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57702_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MFN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MFN1. Alternate names: DKFZp762F247, FLJ20693, MGC41806, hfzo1, hfzo2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-353