Learn More
Aviva Systems Biology Corp MFN2 Antibody - N-terminal region (ARP42419_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42419_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MFN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MFN2. Alternate names: CMT2A, CMT2A2, CPRP1, HSG, KIAA0214, MARFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58221