Learn More
Aviva Systems Biology Corp Mfng Antibody - middle region (ARP45395_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45395_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Mfng. It was validated on Western Blot. . Alternate names: AW546563ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-196