Learn More
Aviva Systems Biology Corp MFRP Antibody - N-terminal region (ARP41281_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41281_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MFRP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MFRP. Alternate names: FLJ30570, NNO2, rd6, RD6, MCOP5ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57793