Learn More
Aviva Systems Biology Corp MFSD1 Antibody - middle region (ARP49741_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49741_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MFSD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MFSD1. Alternate names: FLJ14153, UG0581B09, SMAP4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60165