Learn More
Aviva Systems Biology Corp Mfsd11 Antibody - C-terminal region (ARP49814_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49814_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Mfsd11. It was validated on Western Blot. . Alternate names: 2600014M03Rik, MGC113819ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-453