Learn More
Aviva Systems Biology Corp MFSD3 Antibody - C-terminal region (ARP51707_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51707_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MFSD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MFSD3. Alternate names: -ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-53664