Learn More
Medchemexpress LLC HY-18964 5mg Medchemexpress, MG-101 CAS:110044-82-1 Purity:>98%
SDP
Catalog No. 502082101
Encompass_Preferred
Medchemexpress, HY-18964 5mg MG-101 CAS:110044-82-1 MG-101 is a potent inhibitor of cysteine proteases which inhibits calpain I, calpain II, cathepsin B and cathepsin L with Kis of 190, 220, 150 and 500 pM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2101 Supplier Medchemexpress LLC Supplier No. HY189645MG
Medchemexpress, HY-18964 5mg MG-101 CAS:110044-82-1 MG-101 is a potent inhibitor of cysteine proteases which inhibits calpain I, calpain II, cathepsin B and cathepsin L with Kis of 190, 220, 150 and 500 pM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.