Learn More
Selleck Chemical LLC MG-101
SDP
Catalog No. 501364726
Encompass_Preferred
MG-101 (ALLN Calpain inhibitor-1 Ac-LLnL-CHO) is a cell-permeable and potent inhibitor of cysteine proteases including calpains and lysosomal cathepsins
Catalog No. 50-136-4726 Supplier Selleck Chemical LLC Supplier No. S738625MG
Add to Cart
Add to Cart
MG-101 (ALLN Calpain inhibitor-1 Ac-LLnL-CHO) is a cell-permeable and potent inhibitor of cysteine proteases including calpains and lysosomal cathepsins