Learn More
Selleck Chemical LLC MG-101 (ALLN) 5mg 110044-82-1 Calpain inhibitor-1
SDP
Catalog No. 501364725
Encompass_Preferred
MG-101 (ALLN, Calpain inhibitor-1) is a cell-permeable and potent inhibitor of cysteine proteases including calpains and lysosomal cathepsins. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4725 Supplier Selleck Chemical LLC Supplier No. S73865MG
MG-101 (ALLN, Calpain inhibitor-1) is a cell-permeable and potent inhibitor of cysteine proteases including calpains and lysosomal cathepsins. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.