Learn More
Apexbio Technology LLC MG-115, 25mg. Cas: 133407-86-0 MFCD: MFCD00671410. Potent reversible proteasome inhibitor
SDP
Supplier:  Apexbio Technology LLC A261225
Encompass_Preferred
MG-115 (Z-Leu-Leu-Nva-H) is a potent, reversible peptide aldehyde inhibitor of proteasome chymotrypsin-like and caspase-like activities. It induces p53 dependent apoptosis. Blockade of proteasomal degradation by MG115 can activate autophagy. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4755