Learn More
Medchemexpress LLC HY-13259 25mg Medchemexpress, MG-132 CAS:133407-82-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1325925MG
Encompass_Preferred
Medchemexpress, HY-13259 25mg MG-132 CAS:133407-82-6 MG-132 (Z-Leu-Leu-Leu-al) is a potent proteasome and calpain inhibitor with IC50s of 100 nM and 1.2 μM, respectively. MG-132 effectively blocks the proteolytic activity of the 26S proteasome complex. MG-132, a peptide aldehyde, also is an autophagy activator[2][3]. MG-132 also induces apoptosis[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8973