Learn More
Enzo Life Sciences MG-132 (25 mg) CAS: 133407-82-6
SDP
Catalog No. NC9995990
Encompass_Preferred
Key inhibitor for use in proteasome research. Alternative Name: Z-LLL-CHO. Formula: C26H41N3O5. MW: 475.6. CAS: 133407-82-6. Purity: ≥98%. Sequence Z-Leu-Leu-Leu-CHO. Appearance: White solid. Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: -80°C. Use/Stability: Solutions are stable for up to one week if stored at -20°C. Solutions are stable for up to two months if stored at -80°C.
Catalog No. NC9995990 Supplier Enzo Life Sciences Supplier No. BMLPI1020025
Key inhibitor for use in proteasome research. Alternative Name: Z-LLL-CHO. Formula: C26H41N3O5. MW: 475.6. CAS: 133407-82-6. Purity: ≥98%. Sequence Z-Leu-Leu-Leu-CHO. Appearance: White solid. Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: -80°C. Use/Stability: Solutions are stable for up to one week if stored at -20°C. Solutions are stable for up to two months if stored at -80°C.