Learn More
Medchemexpress LLC HY-13259 100mg , MG-132 CAS:133407-82-6 Purity:98%
SDP
Catalog No. 501492668
Encompass_Preferred
Medchemexpress, HY-13259 100mg MG-132 CAS:133407-82-6 Formula:C26H41N3O5 IC50: 24.2 nM (chymotrypsin-like activity) Purity:98% MG-132 is a potent, non-specific 20S proteasome inhibitor, with IC 50 of 24.2 nM for the β5 chymotrypsin -like active site. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2668 Supplier Medchemexpress LLC Supplier No. HY13259100MG
Medchemexpress, HY-13259 100mg MG-132 CAS:133407-82-6 Formula:C26H41N3O5 IC50: 24.2 nM (chymotrypsin-like activity) Purity:98% MG-132 is a potent, non-specific 20S proteasome inhibitor, with IC 50 of 24.2 nM for the β5 chymotrypsin -like active site. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.